Sebacoyl dichloride ≥97%

Supplier: BeanTown Chemical

125255-100G 125255-25G 125255-500G
BT125255-100GEA 139.34 USD
BT125255-100G BT125255-25G BT125255-500G
Sebacoyl dichloride ≥97%
Sebacoyl dichloride

CAS: 111-19-3; EC No: 203-843-4; MDL No: MFCD00000770; RTECS: HD8454250 UN No: UN2922; Haz Class: 8 (6.1); Packing Group: II Liquid; Linear Formula: ClCO(CH2)8COCl; Molecular Formula: C10H16Cl2O2; MW: 239.14 Melting Point: -5°; Boiling Point: 168°/12 mmHg; Flash point: 110°C (230°F) Density (g/mL): 1.121; Refractive Index: 1.468 Moisture Sensitive


  • RTECS: HD8454250
  • Refractive index: 1.468
  • Moisture Sensitive
  • Liquid

Formula: C₁₀H₁₆Cl₂O₂
Boiling Pt: 168 °C/12 mmHg
Melting Pt: –5 °C
Density: 1.121 g/mL
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00000770
CAS Number: 111-19-3
EINECS: 203-843-4
UN: 2922
ADR: 8,II

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR