Sodium metavanadate ≥96%

Supplier: BeanTown Chemical

Synonyms: Sodium vanadium trioxide

125240-1KG 125240-250G 125240-25G
BT125240-1KGEA 398.64 USD
BT125240-1KG BT125240-250G BT125240-25G
Sodium metavanadate ≥96%
Sodium metavanadate

CAS: 13718-26-8; EC No: 237-272-7; MDL No: MFCD00011125; RTECS: YW1050000 UN No: UN3285; Haz Class: 6.1; Packing Group: III Powder/Chunks; Linear Formula: NaVO3; MW: 121.93 Melting Point: 630° Hygroscopic


  • RTECS: YW1050000
  • Hygroscopic
  • Powder/Chunks

Formula: NaO₃V
MW: 121.93 g/mol
Melting Pt: 630 °C
MDL Number: MFCD00011125
CAS Number: 13718-26-8
EINECS: 237-272-7
UN: 3285
ADR: 6.1,III

Order Now

Specification Test Results

Assay 96%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR