tert-Butoxybis(dimethylamino)methane ≥90%, Technical Grade

Supplier: BeanTown Chemical

Synonyms: Bredereck's reagent, 1-tert-Butoxy-N,N,N',N'-tetramethylmethylenenediamine, [(tert-butoxy)(dimethylamino)methyl]dimethylamine, tert-Butoxy bis(dimethylamino)methane

125220-1G 125220-25G 125220-5G
BT125220-1GEA 45 USD
BT125220-1G BT125220-25G BT125220-5G
tert-Butoxybis(dimethylamino)methane ≥90%, Technical Grade
tert-Butoxybis(dimethylamino)methane

CAS: 5815-08-7; EC No: 227-383-9; MDL No: MFCD00042858 UN No: UN1993; Haz Class: 3; Packing Group: III Liquid; Linear Formula: (CH3)3COCH[N(CH3)2]2; Molecular Formula: C9H22N2O; MW: 174.29 Boiling Point: 50-55°/15 mmHg; Flash point: 41°C (106°F) Density (g/mL): 0.844; Refractive Index: 1.422


  • Refractive index: 1.422
  • Liquid

Formula: C₉H₂₂N₂O
MW: 174.288 g/mol
Boiling Pt: 50…55 °C/15 mmHg
Density: 0.844 g/mL
Flash Pt: 41 °C (106 °F)
MDL Number: MFCD00042858
CAS Number: 5815-08-7
EINECS: 227-383-9
ADR: 3,III

Order Now

Specification Test Results

Assay 90%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR