4-(Trifluoromethyl)phenyl isocyanate ≥98%

Supplier: BeanTown Chemical

Synonyms: α,α,α-Trifluoro-p-tolyl isocyanate, 1-Isocyanato-4-(trifluoromethyl)benzene, 4-(Trifluoromethyl)phenylisocyanate, p-Trifluoromethylphenylisocyanate

125205-1G 125205-5G
BT125205-1GEA 31.21 USD
BT125205-1G BT125205-5G
4-(Trifluoromethyl)phenyl isocyanate ≥98%
4-(Trifluoromethyl)phenyl isocyanate

CAS: 1548-13-6; EC No: 216-284-6; MDL No: MFCD00002028 UN No: UN2285; Haz Class: 6.1; Packing Group: II Liquid; Molecular Formula: C8H4F3NO; MW: 187.12 Boiling Point: 58-59°/10 mmHg; Flash point: 69°C (156°F) Density (g/mL): 1.31; Refractive Index: 1.474 Moisture Sensitive


  • Refractive index: 1.474
  • Moisture Sensitive
  • Liquid

Formula: C₈H₄F₃NO
MW: 187.12 g/mol
Boiling Pt: 58…59 °C/10 mmHg
Density: 1.31 g/mL
Flash Pt: 69 °C (156 °F)
MDL Number: MFCD00002028
CAS Number: 1548-13-6
EINECS: 216-284-6
UN: 2285
ADR: 6.1,II

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR