Graphite natural 99,5 Trace metals basis, flake -325 mesh

Supplier: BeanTown Chemical

125185-1KG 125185-250G 125185-50G
BT125185-1KGEA 111.86 USD
BT125185-1KG BT125185-250G BT125185-50G
Graphite natural 99,5 Trace metals basis, flake -325 mesh
Graphite

CAS: 7782-42-5; EC No: 231-955-3; MDL No: MFCD00144065; RTECS: MD9659600 UN No: UN1325; Haz Class: 4.1; Packing Group: III Solid; Molecular Formula: C; MW: 12.011


  • RTECS: MD9659600
  • Solid

Formula: C
MW: 12.01 g/mol
Melting Pt: 3827
MDL Number: MFCD00144065
CAS Number: 7782-42-5
EINECS: 231-955-3
ADR: 4.1 III
Merck Index: 12,04560

Order Now

Specification Test Results

Assay 99.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR