Cadmium(II) sulphate hydrate ≥99.995%

Supplier: BeanTown Chemical

125180-10G 125180-50G
BT125180-10GEA 68 USD
BT125180-10G BT125180-50G
Cadmium(II) sulphate hydrate ≥99.995%
Cadmium(II) sulphate hydrate

CAS: 15244-35-6; EC No: 233-331-6; MDL No: MFCD00151655 UN No: UN2570; Haz Class: 6.1; Packing Group: III Crystalline; Molecular Formula: 3CdSO4·xH2O ; MW: 208.47 (anhydrous)


  • Crystalline

Formula: CdO₄S·H₂O
MW: 226.49 g/mol
MDL Number: MFCD00151655
CAS Number: 15244-35-6
EINECS: 233-331-6
UN: 2570
ADR: 6.1 III

Order Now

Specification Test Results

Assay 99.995%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR