Bismuth(III) bromide ≥99%
Supplier: BeanTown Chemical
125160-100G
125160-25G
BT125160-100GEA
324
USD
BT125160-100G
BT125160-25G
Bismuth(III) bromide ≥99%
Bismuth(III) bromide
CAS: 7787-58-8; EC No: 232-121-1; MDL No: MFCD00014195
UN No: UN3260; Haz Class: 8; Packing Group: III
Powder; Molecular Formula: BiBr3; MW: 448.69
Density (g/mL): 5.7
Hygroscopic
- Hygroscopic
- Powder
Formula:
BiBr₃ MW: 448.69 g/mol Density: 5.7 g/mL |
MDL Number:
MFCD00014195 CAS Number: 7787-58-8 EINECS: 232-121-1 UN: 3260 ADR: 8,III |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...