Vanadium(III) chloride ≥99%

Supplier: BeanTown Chemical

125125-10G 125125-50G
BT125125-10GEA 106.13 USD
BT125125-10G BT125125-50G
Vanadium(III) chloride ≥99%
Vanadium(III) chloride

CAS: 7718-98-1; EC No: 231-744-6; MDL No: MFCD00011454 UN No: UN2475; Haz Class: 8; Packing Group: III -4 Mesh Powder; Linear Formula: VCl3 ; MW: 157.30 Melting Point: 300° Density (g/mL): 3 Air Sensitive, Light Sensitive, Moisture Sensitive


  • Air Sensitive, Light Sensitive, Moisture Sensitive
  • -4 Mesh Powder

Formula: Cl₃V
MW: 157.3
Melting Pt: >300 °C
Density: 3 g/mL
MDL Number: MFCD00011454
CAS Number: 7718-98-1
EINECS: 231-744-6
UN: 2475
ADR: 8 III

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR