Indium arsenide ≥99%
Supplier: BeanTown Chemical
125085-1G
125085-5G
BT125085-1GEA
38
USD
BT125085-1G
BT125085-5G
Indium arsenide ≥99%
Indium arsenide
CAS: 1303-11-3; EC No: 215-115-3; MDL No: MFCD00016144
UN No: UN1557; Haz Class: 6.1; Packing Group: III
5mm & Down Pieces; Linear Formula: InAs; MW: 189.74
Melting Point: 936°
- 5mm & Down Pieces
Formula:
AsIn Melting Pt: 936 °C |
MDL Number:
MFCD00016144 CAS Number: 1303-11-3 EINECS: 215-115-3 UN: 1557 ADR: 6.1,III |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...