Potassium disulphate mixture of potassium pyrosulfate and potassium bisulfate ACS, fused
Supplier: BeanTown Chemical
Synonyms:
Potassium disulphate (Potassium pyrosulphate), Dipotassium disulphate, Potassium pyrosulphate
125075-100G
125075-500G
BT125075-100GEA
213.41
USD
BT125075-100G
BT125075-500G
Potassium disulphate mixture of potassium pyrosulfate and potassium bisulfate ACS, fused
Potassium disulphate
MDL No: MFCD00066385
UN No: UN2509; Haz Class: 8; Packing Group: II
Fused Powder; Linear Formula: KHSO4 + K2S2O7
Melting Point: 214°
Hygroscopic
- Hygroscopic
- Fused Powder
Formula:
K₂O₇S₂ MW: 254.32 g/mol Melting Pt: 325 |
MDL Number:
MFCD00011385 CAS Number: 7790-62-7 ADR: 8 II Merck Index: 13,07749 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...