2-Methyl-2-butanol ≥98%
Supplier: BeanTown Chemical
CAS: 75-85-4; EC No: 200-908-9; MDL No: MFCD00004478; RTECS: SC0175000
UN No: UN1105; Haz Class: 3; Packing Group: II
Liquid; Linear Formula: CH3CH2C(CH3)2OH; Molecular Formula: C5H12O; MW: 88.15
Melting Point: -12°; Boiling Point: 102°; Flash point: 20°C (68°F)
Density (g/mL): 0.805; Refractive Index: 1.405
- RTECS: SC0175000
- Refractive index: 1.405
- Liquid
Formula:
C₅H₁₂O Boiling Pt: 102 °C Melting Pt: –12 °C Density: 0.805 g/mL Flash Pt: 20 °C (68 °F) |
MDL Number:
MFCD00004478 CAS Number: 75-85-4 EINECS: 200-908-9 UN: 1105 ADR: 3,II |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...