N,N-Diisopropylcarbamoyl chloride ≥98%

Supplier: BeanTown Chemical

Synonyms: Diisopropylcarbamoyl Chloride

125035-25G 125035-5G
BT125035-25GEA 188 USD
BT125035-25G BT125035-5G
N,N-Diisopropylcarbamoyl chloride ≥98%
N,N-Diisopropylcarbamoyl chloride

CAS: 19009-39-3; EC No: 242-743-5; MDL No: MFCD00015013 UN No: UN3261; Haz Class: 8; Packing Group: II Crystalline; Linear Formula: [(CH3)2CH]2NCOCl; Molecular Formula: C7H14ClNO; MW: 163.65 Melting Point: 57-59°; Boiling Point: 90-93°/15 mmHg Moisture Sensitive


  • Moisture Sensitive
  • Crystalline

Formula: C₇H₁₄ClNO
Boiling Pt: 90…93 °C/15 mmHg
Melting Pt: 57…59 °C
MDL Number: MFCD00015013
CAS Number: 19009-39-3
EINECS: 242-743-5
ADR: 8,II

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR