Lead(II) citrate trihydrate ≥99%

Supplier: BeanTown Chemical

Synonyms: Lead citrate, trihydrate

125020-100G 125020-500G
BT125020-100GEA 132 USD
BT125020-100G BT125020-500G
Lead(II) citrate trihydrate ≥99%
Lead(II) citrate trihydrate

CAS: 6107-83-1; EC No: 208-141-1; MDL No: MFCD00150456 UN No: UN2291; Haz Class: 6.1; Packing Group: III Powder; Molecular Formula: C12H10O14Pb3·3H2O; MW: 1053.82


  • Powder

Formula: C₁₂H₁₆O₁₇Pb₃
MDL Number: MFCD00150456
CAS Number: 6107-83-1
EINECS: 208-141-1
ADR: 6.1,III

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR