Barium isopropoxide ≥99.5% (trace metals basis)

Supplier: BeanTown Chemical

Synonyms: Barium i-propoxide

124985-1G 124985-25G 124985-5G
BT124985-1GEA 45 USD
BT124985-1G BT124985-25G BT124985-5G
Barium isopropoxide ≥99.5% (trace metals basis)
Barium isopropoxide

CAS: 24363-37-9; MDL No: MFCD00050486 UN No: UN3205; Haz Class: 4.2; Packing Group: III Powder/Lump; Linear Formula: Ba(OCH(CH3)2)2; MW: 255.50 Melting Point: 200° (decomposes) Moisture Sensitive


  • Moisture Sensitive
  • Powder/Lump

Formula: C₆H₁₄BaO₂
MW: 255.501 g/mol
Melting Pt: 200 °C (dec.)
MDL Number: MFCD00050486
CAS Number: 24363-37-9
UN: 3205
ADR: 4.2,III

Order Now

Specification Test Results

Assay 99.5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR