o-Tolidine ≥98%

Supplier: BeanTown Chemical

Synonyms: 4,4'-Bi-o-toluidine

124975-100G 124975-25G
BT124975-100GEA 235.42 USD
BT124975-100G BT124975-25G
o-Tolidine ≥98%
o-Tolidine

CAS: 119-93-7; EC No: 204-358-0; MDL No: MFCD00014773; RTECS: DD1225000 UN No: UN3077; Haz Class: 9; Packing Group: III Powder; Molecular Formula: C14H16N2; MW: 212.30 Melting Point: 128-131°; Boiling Point: 200°; Flash point: 100°C (212°F) Air Sensitive, Light Sensitive


  • RTECS: DD1225000
  • Air Sensitive, Light Sensitive
  • Powder

Formula: [-C₆H₃(CH₃)-₄-NH₂]₂
MW: 212.29 g/mol
Boiling Pt: 200 °C
Melting Pt: 128…131 °C
Flash Pt: 100 °C (212 °F)
MDL Number: MFCD00014773
CAS Number: 119-93-7
EINECS: 204-358-0
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR