Gadolinium(III) nitrate hydrate 99,9
Supplier: BeanTown Chemical
Synonyms:
GADOLINIUM NITRATE, Gadolinium(III) nitrate hydrate
124970-100G
124970-25G
BT124970-100GEA
117
USD
BT124970-100G
BT124970-25G
Gadolinium(III) nitrate hydrate 99,9
Gadolinium(III) nitrate hydrate
CAS: 94219-55-3; MDL No: MFCD00011023
UN No: UN1477; Haz Class: 5.1; Packing Group: III
Crystalline; Linear Formula: Gd(NO3)3·xH2O; MW: 343.26 (anhydrous)
Melting Point: 91°
Hygroscopic
- Hygroscopic
- Crystalline
Formula:
Gd(NO₃)₃·xH₂O MW: 343.26 g/mol Melting Pt: 91 °C |
MDL Number:
MFCD00149730 CAS Number: 94219-55-3 ADR: 5.1 III |
Specification Test Results
Assay | 99.9% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...