Mercury selenide ≥99.9% (trace metals basis)

Supplier: BeanTown Chemical

124950-1G 124950-5G
BT124950-1GEA 37 USD
BT124950-1G BT124950-5G
Mercury selenide ≥99.9% (trace metals basis)
Mercury selenide

CAS: 20601-83-6; EC No: 243-910-5; MDL No: MFCD00049743 UN No: UN2025; Haz Class: 6.1; Packing Group: II -100 Mesh Powder; Molecular Formula: HgSe; MW: 279.55 Density (g/mL): 8.21


  • -100 Mesh Powder

Formula: HgSe
MW: 279.55 g/mol
Density: 8.21 g/mL
MDL Number: MFCD00049743
CAS Number: 20601-83-6
EINECS: 243-910-5
UN: 2025
ADR: 6.1,II

Order Now

Specification Test Results

Assay 99.9%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR