Toluene-p-sulfonic anhydride ≥97%

Supplier: BeanTown Chemical

Synonyms: 4-Methylbenzenesulphonic anhydride, p-Toluenesulphonic anhydride

124925-25G 124925-5G
BT124925-25GEA 153 USD
BT124925-25G BT124925-5G
Toluene-p-sulfonic anhydride ≥97%
Toluene-p-sulphonic anhydride

CAS: 4124-41-8; EC No: 223-926-9; MDL No: MFCD00008548 UN No: UN3261; Haz Class: 8; Packing Group: III Powder; Linear Formula: (CH3C6H4SO2)2O; Molecular Formula: C14H14O5S2; MW: 326.38 Melting Point: 121-127° Moisture Sensitive


  • Moisture Sensitive
  • Powder

Formula: C₁₄H₁₄O₅S₂
MW: 326.39 g/mol
Melting Pt: 121…127 °C
MDL Number: MFCD00008548
CAS Number: 4124-41-8
EINECS: 223-926-9
UN: 3261
ADR: 8,III

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR