Potassium tetraoxalate dihydrate ≥98%

Supplier: BeanTown Chemical

Synonyms: Oxalic acid, potassium salt dihydrate, Potassium trihydrogen dioxalate dihydrate

124880-1KG 124880-250G 124880-5KG
BT124880-1KGEA 137.82 USD
BT124880-1KG BT124880-250G BT124880-5KG
Potassium tetraoxalate dihydrate ≥98%
Potassium tetraoxalate dihydrate

CAS: 6100-20-5; EC No: 204-874-6; MDL No: MFCD00150443 UN No: UN3288; Haz Class: 6.1; Packing Group: III Crystalline; Linear Formula: KH3(C2O4)2·2H2O; MW: 254.19 Density (g/mL): 1.84


  • Crystalline

Formula: C₄H₃KO₈·2H₂O
MW: 254.19 g/mol (218.16 anhy)
Density: 1.84 g/mL
MDL Number: MFCD00150443
CAS Number: 6100-20-5
EINECS: 204-874-6
UN: 3288
ADR: 6.1 III

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR