Bis(cyclopentadienyl)cobalt

Supplier: BeanTown Chemical

Synonyms: Cobaltocene, Bis(cyclopentadienyl)cobalt(II)

124815-1G 124815-25G 124815-5G
BT124815-1GEA 55.22 USD
BT124815-1G BT124815-25G BT124815-5G
Bis(cyclopentadienyl)cobalt
Bis(cyclopentadienyl)cobalt

CAS: 1277-43-6; EC No: 215-061-0; MDL No: MFCD00013749 UN No: UN3181; Haz Class: 4.1; Packing Group: II Powder; Molecular Formula: C10H10Co; MW: 189.12 Melting Point: 176-180° (decomposes) Air Sensitive, Moisture Sensitive


  • Air Sensitive, Moisture Sensitive
  • Powder

Formula: C₁₀H₁₀Co
MW: 189.12 g/mol
Melting Pt: 176…180 °C (dec.)
MDL Number: MFCD00013749
CAS Number: 1277-43-6
EINECS: 215-061-0
UN: 3181
ADR: 4.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR