Cobalt(II,III) oxide ≥99%
Supplier: BeanTown Chemical
Synonyms:
Tricobalt tetraoxide, Cobaltic tetraoxide
124805-100G
124805-25G
124805-500G
BT124805-100GEA
356
USD
BT124805-100G
BT124805-25G
BT124805-500G
Cobalt(II,III) oxide ≥99%
Cobalt(II,III) oxide
CAS: 1308-06-1; EC No: 215-157-2; MDL No: MFCD00010939; RTECS: GG2500000
UN No: UN3077; Haz Class: 9; Packing Group: III
50-80nm APS Powder, S.A. 10 m2/g; Molecular Formula: Co3O4; MW: 240.80
Melting Point: 895° (decomposes)
Density (g/mL): 6.11
- RTECS: GG2500000
- 50-80nm APS Powder, S.A. 10 m²/g
Formula:
Co₃O₄ MW: 240.8 g/mol Melting Pt: 895 (dec.) Density: 6.11 g/mL |
MDL Number:
MFCD00010939 CAS Number: 1308-06-1 EINECS: 215-157-2 ADR: 9 III Merck Index: 13,02454 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...