α,4-Dibromotoluene ≥98%

Supplier: BeanTown Chemical

Synonyms: 4-Bromobenzyl bromide, 1-Bromo-4-(bromomethyl)benzene

124800-100G 124800-25G
BT124800-100GEA 115 USD
BT124800-100G BT124800-25G
α,4-Dibromotoluene ≥98%
α,4-Dibromotoluene

CAS: 589-15-1; EC No: 209-636-5; MDL No: MFCD00000179 UN No: UN1759; Haz Class: 8; Packing Group: II Crystalline; Molecular Formula: C7H6Br2; MW: 249.93 Melting Point: 62-64°


  • Crystalline

Formula: C₇H₆Br₂
MW: 249.93 g/mol
Melting Pt: 62…64 °C
MDL Number: MFCD00000179
CAS Number: 589-15-1
EINECS: 209-636-5
UN: 1759
ADR: 6.1,II

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR