Osmium tetraoxide ≥99.8%

Supplier: BeanTown Chemical

Synonyms: Osmium (VIII) oxide, Osmic acid anhydride, Osmic acid

124780-1G 124780-500MG
BT124780-1GEA 164 USD
BT124780-1G BT124780-500MG
Osmium tetraoxide ≥99.8%
Osmium tetraoxide

CAS: 20816-12-0; EC No: 244-058-7; MDL No: MFCD00011150 UN No: UN2471; Haz Class: 6.1; Packing Group: I Crystalline; Linear Formula: OsO4; MW: 254.20 Melting Point: 40°; Boiling Point: 130° Density (g/mL): 4.9


  • Crystalline

Formula: O₄Os
Boiling Pt: 130 °C
Melting Pt: 40 °C
Density: 4.9 g/mL
MDL Number: MFCD00011150
CAS Number: 20816-12-0
EINECS: 244-058-7
UN: 2471
ADR: 6.1,I

Order Now

Specification Test Results

Assay 99.8%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR