Rhenium ≥99.99% (trace metals basis), powder -325 mesh
Supplier: BeanTown Chemical
124730-10G
124730-2G
124730-500MG
BT124730-10GEA
485
USD
BT124730-10G
BT124730-2G
BT124730-500MG
Rhenium ≥99.99% (trace metals basis), powder -325 mesh
Rhenium
CAS: 7440-15-5; EC No: 231-124-5; MDL No: MFCD00011195; RTECS: VI0780000
UN No: UN3089; Haz Class: 4.1; Packing Group: III
Powder; Molecular Formula: Re; MW: 186.21
Melting Point: 3180°; Boiling Point: 5596°
Density (g/mL): 21.02
- RTECS: VI0780000
- Powder
Formula:
Re MW: 186.21 g/mol Boiling Pt: 5596 °C Melting Pt: 3180 °C Density: 21.02 g/mL |
MDL Number:
MFCD00011195 CAS Number: 7440-15-5 EINECS: 231-124-5 UN: 3089 ADR: 4.1,III |
Specification Test Results
Assay | 99.99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...