Ammonium iodate ≥99%

Supplier: BeanTown Chemical

Synonyms: Ammonium Iodate, Iodic acid, ammonium salt

124710-250G 124710-50G
BT124710-250GEA 199 USD
BT124710-250G BT124710-50G
Ammonium iodate ≥99%
Ammonium iodate

CAS: 13446-09-8; EC No: 236-592-4; MDL No: MFCD00049684 UN No: UN1479; Haz Class: 5.1; Packing Group: II Crystalline; Linear Formula: NH4IO3; MW: 192.94 Light Sensitive, Moisture Sensitive


  • Light Sensitive, Moisture Sensitive
  • Crystalline

Formula: H₄INO₃
MW: 192.94
Melting Pt: 150
Density: 3.407
MDL Number: MFCD00049684
CAS Number: 13446-09-8
EINECS: 236-592-4
UN: 1479
ADR: 5.1 II

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR