Copper(II) chloride hydrate ≥99.999% (trace metals basis)
Supplier: BeanTown Chemical
|
Danger
|
CAS: 10125-13-0; EC No: 231-210-2; MDL No: MFCD00149792; RTECS: GL7030000
UN No: UN2802; Haz Class: 8; Packing Group: III
Crystalline Aggregates; Linear Formula: CuCl2·xH2O; MW: 134.45 (anhydrous)
Melting Point: 100° (decomposes)
Air Sensitive , Hygroscopic
- RTECS: GL7030000
- Air Sensitive, Hygroscopic
- Crystalline Aggregates
Formula:
Cl₂Cu·xH₂O Melting Pt: 100 °C (dec.) |
CAS Number:
10125-13-0 EINECS: 231-210-2 UN: 2802 ADR: 8,III Merck Index: 14,2633 |
Specification Test Results
Assay | 99.999% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...