Sulfur trioxide pyridine complex ≥98% active SO₃ ∼ 48-50%

Supplier: BeanTown Chemical

124630-10G 124630-250G 124630-50G
BT124630-10GEA 22.97 USD
BT124630-10G BT124630-250G BT124630-50G
Sulfur trioxide pyridine complex ≥98% active SO₃ ∼ 48-50%
Sulphur trioxide pyridine complex

CAS: 26412-87-3; EC No: 247-683-3; MDL No: MFCD00012437 UN No: UN3261; Haz Class: 8; Packing Group: II Powder; Molecular Formula: C5H5NO3S; MW: 159.16 Melting Point: 155-165° Moisture Sensitive


  • Moisture Sensitive
  • Powder

Formula: C₅H₅NO₃S
MW: 159.16 g/mol
Melting Pt: 155…165 °C
MDL Number: MFCD00012437
CAS Number: 26412-87-3
EINECS: 247-683-3
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR