Cyclohexene-1-carbonitrile ≥98%

Supplier: BeanTown Chemical

Synonyms: 1-Cyclohexene-1-carbonitrile, Cyclohex-1-enecarbonitrile

124585-1G 124585-5G
BT124585-1GEA 26 USD
BT124585-1G BT124585-5G
Cyclohexene-1-carbonitrile ≥98%
Cyclohexene-1-carbonitrile

CAS: 1855-63-6; EC No: 217-454-2; MDL No: MFCD00013772 UN No: UN3276; Haz Class: 6.1; Packing Group: III Liquid; Molecular Formula: C7H9N; MW: 107.15 Flash point: 83°C (181°F) Density (g/mL): 0.946; Refractive Index: 1.482


  • Refractive index: 1.482
  • Liquid

Formula: C₇H₉N
MW: 107.16 g/mol
Density: 0.946 g/mL
Flash Pt: 83 °C (181 °F)
MDL Number: MFCD00013772
CAS Number: 1855-63-6
EINECS: 217-454-2
UN: 3276
ADR: 6.1,III

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR