Hydrobromic acid 47.0 - 49.0% ACS
Supplier: BeanTown Chemical
124575-1L
124575-250ML
124575-4X1L
BT124575-1LEA
162.34
USD
BT124575-1L
BT124575-250ML
BT124575-4X1L
Hydrobromic acid 47.0 - 49.0% ACS
Hydrobromic acid
CAS: 10035-10-6; EC No: 233-113-0; MDL No: MFCD00011323; RTECS: MW3850000
UN No: UN1788; Haz Class: 8; Packing Group: II
Liquid; Molecular Formula: HBr; MW: 80.92
Density (g/mL): 1.490
Air Sensitive
- RTECS: MW3850000
- Air Sensitive
- Liquid
Formula:
HBr Density: 1.490 g/mL |
MDL Number:
MFCD00011323 CAS Number: 10035-10-6 EINECS: 233-113-0 UN: 1788 ADR: 8,II |
Specification Test Results
Assay | 47.0-49.0% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...