Mercury(II) trifluoromethanesulfonate ≥98%
Supplier: BeanTown Chemical
Synonyms:
Mercury(II) trifluoromethanesulphonate, Mercury triflate
124540-1G
124540-25G
124540-5G
BT124540-1GEA
44
USD
BT124540-1G
BT124540-25G
BT124540-5G
Mercury(II) trifluoromethanesulfonate ≥98%
Mercury(II) trifluoromethanesulphonate
CAS: 49540-00-3; MDL No: MFCD00144746
UN No: UN2025; Haz Class: 6.1; Packing Group: II
Powder; Linear Formula: Hg(O3SCF3)2; Molecular Formula: C2F6HgO6S2 ; MW: 498.72
Melting Point:
- Moisture Sensitive
- Powder
Formula:
C₂F₆HgO₆S₂ MW: 498.73 g/mol Melting Pt: >350 °C |
MDL Number:
MFCD00144746 CAS Number: 49540-00-3 UN: 2025 ADR: 6.1,II |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...