2,6-Toluenediisocyanate ≥97%

Supplier: BeanTown Chemical

Synonyms: 2,6-TDI, 2-Methyl-m-phenylene diisocyanate, Tolylene 2,6-diisocyanate

124485-1G 124485-5G
BT124485-1GEA 42 USD
BT124485-1G BT124485-5G
2,6-Toluenediisocyanate ≥97%
2,6-Toluenediisocyanate

CAS: 91-08-7; EC No: 202-039-0; MDL No: MFCD00002010; RTECS: CZ6310000 UN No: UN2078; Haz Class: 6.1; Packing Group: II Liquid; Molecular Formula: C9H6N2O2; MW: 174.16 Melting Point: 20-22°; Boiling Point: 129-133°/18 mmHg; Flash point: 110°C (230°F) Density (g/mL): 1.225; Refractive Index: 1.571 Light Sensitive, Moisture Sensitive


  • RTECS: CZ6310000
  • Refractive index: 1.571
  • Light Sensitive, Moisture Sensitive
  • Liquid

Formula: C₉H₆N₂O₂
MW: 174.16 g/mol
Boiling Pt: 129…133 °C/18 mmHg
Melting Pt: 20…22 °C
Density: 1.225 g/mL
Flash Pt: 110 °C (230 °F)
MDL Number: MFCD00002010
CAS Number: 91-08-7
EINECS: 202-039-0
UN: 2078
ADR: 6.1,II

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR