Succinonitrile ≥98%

Supplier: BeanTown Chemical

Synonyms: Butanedinitrile, Succinodinitrile

124430-100G 124430-500G
BT124430-100GEA 49.81 USD
BT124430-100G BT124430-500G
Succinonitrile ≥98%
Succinonitrile

CAS: 110-61-2; EC No: 203-783-9; MDL No: MFCD00001949; RTECS: WN3850000 UN No: UN3439; Haz Class: 6.1; Packing Group: III Crystalline; Linear Formula: NCCH2CH2CN; Molecular Formula: C4H4N2; MW: 80.09 Melting Point: 50-54°; Boiling Point: 265-267°; Flash point: 110°C (230°F) Density (g/mL): 0.985


  • RTECS: WN3850000
  • Crystalline

Formula: C₄H₄N₂
MW: 80.09 g/mol
Boiling Pt: 265…267 °C
Melting Pt: 50…54 °C
Density: 0.985 g/mL
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00001949
CAS Number: 110-61-2
EINECS: 203-783-9
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR