Silicon ≥99.5% (trace metals basis), crystalline powder -325 mesh

Supplier: BeanTown Chemical

124420-100G 124420-25G 124420-500G
BT124420-100GEA 182 USD
BT124420-100G BT124420-25G BT124420-500G
Silicon ≥99.5% (trace metals basis), crystalline powder -325 mesh
Silicon

CAS: 7440-21-3; EC No: 231-130-8; MDL No: MFCD00085311; RTECS: VW0400000 UN No: UN1346; Haz Class: 4.1; Packing Group: III Powder; Molecular Formula: Si; MW: 28.09 Melting Point: 1410°; Boiling Point: 2355° Density (g/mL): 2.33


  • RTECS: VW0400000
  • Powder

Formula: Si
Boiling Pt: 2355 °C
Melting Pt: 1410 °C
Density: 2.33 g/mL
MDL Number: MFCD00085311
CAS Number: 7440-21-3
EINECS: 231-130-8
UN: 1346
ADR: 4.1,III

Order Now

Specification Test Results

Assay 99.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR