Cadmium(II) bromide, anhydrous ≥98%
Supplier: BeanTown Chemical
124375-100G
124375-25G
BT124375-100GEA
176.32
USD
BT124375-100G
BT124375-25G
Cadmium(II) bromide, anhydrous ≥98%
Cadmium(II) bromide
CAS: 7789-42-6; EC No: 232-165-1; MDL No: MFCD00015996; RTECS: EU9935000
UN No: UN2570; Haz Class: 6.1; Packing Group: III
Powder; Linear Formula: CdBr2 ; MW: 272.22
Hygroscopic
- RTECS: EU9935000
- Hygroscopic
- Powder
Formula:
Br₂Cd |
MDL Number:
MFCD00015996 CAS Number: 7789-42-6 EINECS: 232-165-1 UN: 2570 ADR: 6.1,III |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...