(R)-2-Amino-1-phenylethanol ≥97%

Supplier: BeanTown Chemical

Synonyms: (R)-(-)-2-Amino-1-phenylethanol, (R)-(-)-alpha-(Aminomethyl)benzyl alcohol

124265-1G 124265-5G
BT124265-1GEA 66 USD
BT124265-1G BT124265-5G
(R)-2-Amino-1-phenylethanol ≥97%
(R)-2-Amino-1-phenylethanol

CAS: 2549-14-6; MDL No: MFCD00239406 UN No: UN3259; Haz Class: 8; Packing Group: III Powder; Linear Formula: H2NCH2CH(C6H5)OH; Molecular Formula: C8H11NO ; MW: 137.18 Melting Point: 59-65° Optical Rotation: [α]22/D −39°, c = 2 in ethanol Air Sensitive


  • Optical rotation: [α]22/D −39°, c = 2 in ethanol
  • Air Sensitive
  • Powder

Formula: C₈H₁₁NO
MW: 137.18
Melting Pt: 59…65 °C
MDL Number: MFCD00239406
CAS Number: 2549-14-6
ADR: 8,III

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR