(±)-2-Bromobutyric acid ≥98%
Supplier: BeanTown Chemical
CAS: 80-58-0; EC No: 201-294-5; MDL No: MFCD00004216
UN No: UN3265; Haz Class: 8; Packing Group: II
Liquid; Linear Formula: CH3CH2CHBrCO2H ; Molecular Formula: C4H7BrO2 ; MW: 167.00
Melting Point: -4°; Boiling Point: 99-103°/10 mmHg
Density (g/mL): 1.567; Refractive Index: 1.474
- Refractive index: 1.474
- Liquid
Formula:
H₃CCH₂BrCHCOOH Boiling Pt: 99…103 °C/10 mmHg Melting Pt: –4 °C Density: 1.567 g/mL |
MDL Number:
MFCD00004216 CAS Number: 80-58-0 EINECS: 201-294-5 UN: 3265 ADR: 8,II |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...