Silver phosphate ≥99%
Supplier: BeanTown Chemical
124190-25G
124190-5G
BT124190-25GEA
71
USD
BT124190-25G
BT124190-5G
Silver phosphate ≥99%
Silver phosphate
CAS: 7784-09-0; EC No: 232-049-0; MDL No: MFCD00014148
UN No: UN3077; Haz Class: 9; Packing Group: III
Powder; Linear Formula: Ag3PO4; MW: 418.58
Melting Point: 849°
Density (g/mL): 6.37
Light Sensitive
- Light Sensitive
- Powder
Formula:
Ag₃O₄P MW: 418.58 g/mol Melting Pt: 849 °C Density: 6.37 g/mL |
MDL Number:
MFCD00014148 CAS Number: 7784-09-0 EINECS: 232-049-0 ADR: 9 III |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...