1,8-Bis(dimethylamino)naphthalene ≥98%
Supplier: BeanTown Chemical
Synonyms:
N1,N1,N8,N8-Tetramethylnaphthalene-1,8-diamine
124155-10G
124155-50G
BT124155-10GEA
34
USD
BT124155-10G
BT124155-50G
1,8-Bis(dimethylamino)naphthalene ≥98%
1,8-Bis(dimethylamino)naphthalene
CAS: 20734-58-1; EC No: 244-001-6; MDL No: MFCD00003920
UN No: UN3263; Haz Class: 8; Packing Group: III
Crystalline/Powder; Molecular Formula: C14H18N2; MW: 214.31
Melting Point: 45-49°; Flash point:
- Air Sensitive, Light Sensitive
- Crystalline/Powder
Formula:
C₁₄H₁₈N₂ MW: 214.31 g/mol Melting Pt: 45…49 °C Flash Pt: >110 °C (>230 °F) |
MDL Number:
MFCD00003920 CAS Number: 20734-58-1 EINECS: 244-001-6 UN: 3263 ADR: 8,III |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...