Propiolaldehyde diethyl acetal ≥97%

Supplier: BeanTown Chemical

Synonyms: Propargylaldehyde diethyl acetal, 3,3-Diethoxypropyne, Propioaldehyde diethyl acetal, Propynal diethyl acetal, 3,3-Diethoxy-1-propyne, 3,3-Diethoxyprop-1-yne

124130-1G 124130-25G 124130-5G
BT124130-1GEA 46 USD
BT124130-1G BT124130-25G BT124130-5G
Propiolaldehyde diethyl acetal ≥97%
Propiolaldehyde diethyl acetal

CAS: 10160-87-9; EC No: 233-430-4; MDL No: MFCD00009237 UN No: UN1993; Haz Class: 3; Packing Group: III Liquid; Linear Formula: HC≡CCH(OCH2CH3)2; Molecular Formula: C7H12O2; MW: 128.17 Boiling Point: 138-139.5°; Flash point: 31°C (88°F) Density (g/mL): 0.894; Refractive Index: 1.412 Air Sensitive, Moisture Sensitive


  • Refractive index: 1.412
  • Air Sensitive, Moisture Sensitive
  • Liquid

Formula: C₇H₁₂O₂
MW: 128.17 g/mol
Boiling Pt: 38
Density: 0.894 g/mL
Flash Pt: 31 °C (88 °F)
MDL Number: MFCD00009237
CAS Number: 10160-87-9
EINECS: 233-430-4
ADR: 3 III

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR