3-Nitro-p-cresol ≥98%

Supplier: BeanTown Chemical

Synonyms: 4-Methyl-3-nitrophenol, 1-Hydroxy-4-methyl-3-nitrobenzene

124120-25G 124120-5G
BT124120-25GEA 89 USD
BT124120-25G BT124120-5G
3-Nitro-p-cresol ≥98%
3-Nitro-p-cresol

CAS: 2042-14-0; EC No: 218-044-6; MDL No: MFCD00007244 UN No: UN2446; Haz Class: 6.1; Packing Group: III Powder; Molecular Formula: C7H7NO3; MW: 153.14 Melting Point: 78-81°


  • Powder

Formula: C₇H₇NO₃
MW: 153.14 g/mol
Melting Pt: 78…81 °C
MDL Number: MFCD00007244
CAS Number: 2042-14-0
EINECS: 218-044-6
UN: 2446
ADR: 6.1,III

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR