2-Chloro-6-methylpyridine ≥98%

Supplier: BeanTown Chemical

Synonyms: 6-Chloro-2-picoline

124110-1G 124110-25G 124110-5G
BT124110-1GEA 21 USD
BT124110-1G BT124110-25G BT124110-5G
2-Chloro-6-methylpyridine ≥98%
2-Chloro-6-methylpyridine

CAS: 18368-63-3; EC No: 242-241-6; MDL No: MFCD00006245 UN No: UN2810; Haz Class: 6.1; Packing Group: III Liquid; Molecular Formula: C6H6ClN; MW: 127.57 Boiling Point: 64-68°/10 mmHg; Flash point: 74°C (165°F) Density (g/mL): 1.167; Refractive Index: 1.527


  • Refractive index: 1.527
  • Liquid

Formula: C₆H₆ClN
MW: 127.57 g/mol
Boiling Pt: 64
Density: 1.167 g/mL
Flash Pt: 74 °C (165 °F)
MDL Number: MFCD00006245
CAS Number: 18368-63-3
EINECS: 242-241-6
UN: 2810
ADR: 6.1 III

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR