Cerium ≥99.9% (trace metals basis excluding Ta), rods Ø 12.7 mm

Supplier: BeanTown Chemical

124105-10MM 124105-25MM 124105-50MM
BT124105-10MMEA 151 USD
BT124105-10MM BT124105-25MM BT124105-50MM
Cerium ≥99.9% (trace metals basis excluding Ta), rods Ø 12.7 mm
Cerium

CAS: 7440-45-1; EC No: 231-154-9; MDL No: MFCD00010924 UN No: UN1333; Haz Class: 4.1; Packing Group: II Solid, cast rod, diameter is approximate; Molecular Formula: Ce; MW: 140.12 Melting Point: 795°; Boiling Point: 3443° Density (g/mL): 6.67


  • Solid, cast rod, diameter is approximate

Formula: Ce
MW: 140.12 g/mol
Boiling Pt: 3468
Melting Pt: 795
Density: 6.67 g/mL
MDL Number: MFCD00010924
CAS Number: 7440-45-1
EINECS: 231-154-9
UN: 1333
ADR: 4.1 II

Order Now

Specification Test Results

Assay 99.9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR