1,4-Diazabicyclo[2.2.2]octane ≥98%
Supplier: BeanTown Chemical
Synonyms:
DABCO
124095-100G
124095-25G
124095-500G
BT124095-100GEA
62.35
USD
BT124095-100G
BT124095-25G
BT124095-500G
1,4-Diazabicyclo[2.2.2]octane ≥98%
1,4-Diazabicyclo[2.2.2]octane
CAS: 280-57-9; EC No: 205-999-9; MDL No: MFCD00006689; RTECS: HM0354200
UN No: UN1325; Haz Class: 4.1; Packing Group: III
Solid; Molecular Formula: C6H12N2; MW: 112.17
Melting Point: 156-159°; Flash point: 62°C (144°F)
Density (g/mL): 1.02; Refractive Index: 1.463
Hygroscopic
- RTECS: HM0354200
- Refractive index: 1.463
- Hygroscopic
- Solid
Formula:
C₆H₁₂N₂ MW: 112.17 g/mol Boiling Pt: 174 Melting Pt: 156…159 °C Density: 1.02 g/mL Flash Pt: 62 °C (144 °F) |
MDL Number:
MFCD00006689 CAS Number: 280-57-9 EINECS: 205-999-9 UN: 1325 ADR: 4.1 III Merck Index: 12,09801 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...