Silver carbonate ≥99.5%
Supplier: BeanTown Chemical
124045-100G
124045-25G
BT124045-100GEA
417
USD
BT124045-100G
BT124045-25G
Silver carbonate ≥99.5%
Silver carbonate
CAS: 534-16-7; EC No: 208-590-3; MDL No: MFCD00003403; RTECS: FG0700000
UN No: UN3077; Haz Class: 9; Packing Group: III
Powder; Linear Formula: Ag2CO3; MW: 275.75
Melting Point: 218° (decomposes)
Density (g/mL): 6.08
Light Sensitive
- RTECS: FG0700000
- Light Sensitive
- Powder
Formula:
CAg₂O₃ MW: 275.75 g/mol Melting Pt: 218 °C (dec.) Density: 6.08 g/mL |
MDL Number:
MFCD00003403 CAS Number: 534-16-7 EINECS: 208-590-3 ADR: 9 III Merck Index: 12,08650 |
Specification Test Results
Assay | 99.5% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...