(tert-Butyldimethylsilyl)acetylene ≥98%

Supplier: BeanTown Chemical

124035-1G 124035-25G 124035-5G
BT124035-1GEA 64.94 USD
BT124035-1G BT124035-25G BT124035-5G
(tert-Butyldimethylsilyl)acetylene ≥98%
(tert-Butyldimethylsilyl)acetylene

CAS: 86318-61-8; MDL No: MFCD00191877 UN No: UN1993; Haz Class: 3; Packing Group: II Liquid; Linear Formula: (CH3)3CSi(CH3)2CCH; Molecular Formula: C8H16Si; MW: 140.30 Boiling Point: 116-117°; Flash point: 6°C (43°F) Density (g/mL): 0.751; Refractive Index: 1.451 Moisture Sensitive


  • Refractive index: 1.451
  • Moisture Sensitive
  • Liquid

Formula: C₈H₁₆Si
MW: 140.3 g/mol
Boiling Pt: 116…117 °C
Density: 0.751 g/mL
Flash Pt: 6 °C (43 °F)
MDL Number: MFCD00191877
CAS Number: 86318-61-8
ADR: 3,II

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR