Zinc oxide ≥99.99%
Supplier: BeanTown Chemical
Synonyms:
Zinci oxidum, Zinc (II) oxide
123985-250G
123985-50G
BT123985-250GEA
166.45
USD
BT123985-250G
BT123985-50G
Zinc oxide ≥99.99%
Zinc oxide
CAS: 1314-13-2; EC No: 215-222-5; MDL No: MFCD00011300; RTECS: ZH4810000
UN No: UN3077; Haz Class: 9; Packing Group: III
Powder; Linear Formula: ZnO; MW: 81.39
Melting Point: 1975°
Density (g/mL): 5.61
- RTECS: ZH4810000
- Powder
Formula:
ZnO MW: 81.39 g/mol Melting Pt: 1975 °C Density: 5.61 g/mL |
MDL Number:
MFCD00011300 CAS Number: 1314-13-2 EINECS: 215-222-5 UN: 3077 ADR: 9 III |
Specification Test Results
Assay | 99.99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...