Acetone-[D₆] ≥99.9% (Isotopic)
Supplier: BeanTown Chemical
Synonyms:
Propanone-D6, DMK-D6, Hexadeuteroacetone, Dimethyl ketone-D6, Acetone-[²H₆], Acetone-d6
123980-10G
123980-5X10G
BT123980-10GEA
104.63
USD
BT123980-10G
BT123980-5X10G
Acetone-[D₆] ≥99.9% (Isotopic)
Acetone-[D6]
CAS: 666-52-4; EC No: 211-563-9; MDL No: MFCD00044635
UN No: UN1090; Haz Class: 3; Packing Group: II
Liquid; Linear Formula: CD3COCD3; MW: 64.12
Melting Point: -94°; Boiling Point: 55.5°; Flash point: -19°C (-2°F)
Density (g/mL): 0.872; Refractive Index: 1.355
- Refractive index: 1.355
- Liquid
Formula:
D₃CC(O)CD₃ MW: 64.03 g/mol Boiling Pt: 55.5 °C Melting Pt: –94 °C Density: 0.872 g/mL Flash Pt: –19 °C (–2 °F) |
MDL Number:
MFCD00044635 CAS Number: 666-52-4 EINECS: 211-563-9 UN: 1090 ADR: 3,II |
Specification Test Results
Assay | 99.9% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...