1-Chloro-4-ethynylbenzene ≥98%

Supplier: BeanTown Chemical

Synonyms: (4-Chlorophenyl)acetylene, 1-Ethynyl-4-chlorobenzene, (4-Chlorophenyl)ethyne

123960-1G 123960-5G
BT123960-1GEA 167 USD
BT123960-1G BT123960-5G
1-Chloro-4-ethynylbenzene ≥98%
1-Chloro-4-ethynylbenzene

CAS: 873-73-4; MDL No: MFCD00191917 UN No: UN1325; Haz Class: 4.1; Packing Group: III Solid; Molecular Formula: C8H5Cl; MW: 136.58 Melting Point: 45-47°; Flash point: 10°C (50°F)


  • Solid

Formula: C₈H₅Cl
MW: 136.58 g/mol
Melting Pt: 45…47 °C
Flash Pt: 10 °C (50 °F)
MDL Number: MFCD00191917
CAS Number: 873-73-4
UN: 1325
ADR: 4.1,III

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR