Benzyl isocyanate ≥98%
Supplier: BeanTown Chemical
123925-1G
123925-25G
123925-5G
BT123925-1GEA
26
USD
BT123925-1G
BT123925-25G
BT123925-5G
Benzyl isocyanate ≥98%
Benzyl isocyanate
CAS: 3173-56-6; EC No: 221-640-9; MDL No: MFCD00009701
UN No: UN3080; Haz Class: 6.1 (3); Packing Group: II
Liquid; Linear Formula: C6H5CH2NCO; Molecular Formula: C8H7NO; MW: 133.15
Boiling Point: 101-104°/33 mmHg; Flash point: 45°C (113°F)
Density (g/mL): 1.078; Refractive Index: 1.526
Moisture Sensitive
- Refractive index: 1.526
- Moisture Sensitive
- Liquid
Formula:
C₈H₇NO MW: 133.15 g/mol Boiling Pt: 101…104 °C/33 mmHg Density: 1.078 g/mL Flash Pt: 45 °C (113 °F) |
MDL Number:
MFCD00009701 CAS Number: 3173-56-6 EINECS: 221-640-9 UN: 3080 ADR: 6.1,II |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...