Silver ≥99.9% (trace metals basis), Powder APS 4-7 µm

Supplier: BeanTown Chemical

123870-10G 123870-250G 123870-50G
BT123870-10GEA 58 USD
BT123870-10G BT123870-250G BT123870-50G
Silver ≥99.9% (trace metals basis), Powder APS 4-7 µm
Silver

CAS: 7440-22-4; EC No: 231-131-3; MDL No: MFCD00003397; RTECS: VW3500000 UN No: UN3077; Haz Class: 9; Packing Group: III Powder, S.A. 0.1-0.4 m2/g ; Molecular Formula: Ag; MW: 107.87 Melting Point: 960°; Boiling Point: 2212° Density (g/mL): 10.49


  • RTECS: VW3500000
  • Powder, S.A. 0.1-0.4 m²/g

Formula: Ag
Boiling Pt: 2212 °C
Melting Pt: 960 °C
Density: 10.49 g/mL
MDL Number: MFCD00003397
CAS Number: 7440-22-4
EINECS: 231-131-3
ADR: 9,III

Order Now

Specification Test Results

Assay 99.9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR